Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

611336

Supelco

Iodomethane-13C solution

1% in chloroform-d (99.9 atom % D), trimethyl phosphite 1 %, chromium(III) acetylacetonate 0.2 %, NMR tube size 8 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CH3I
Molecular Weight:
142.93
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

composition

chromium(III) acetylacetonate, 0.2%
trimethyl phosphite, 1%

concentration

1% in chloroform-d (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

8 mm × 8 in.

suitability

suitable for NMR (reference standard)

format

single component solution

mass shift

M+1

SMILES string

[13CH3]I

InChI

1S/CH3I/c1-2/h1H3/i1+1

InChI key

INQOMBQAUSQDDS-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Indirect detection test

Quantity

8 mm O.D. tube contains 1.9 mL.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service