Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

608513

Sigma-Aldrich

DL-Allantoin-5-13C,1-15N

99 atom % 13C, 98 atom % 15N, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC3H615NN3O3
CAS Number:
Molecular Weight:
160.10
UNSPSC Code:
12352106
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
98 atom % 15N

assay

99% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

230 °C (dec.) (lit.)

mass shift

M+2

SMILES string

NC(=O)N[13CH]1[15NH]C(=O)NC1=O

InChI

1S/C4H6N4O3/c5-3(10)6-1-2(9)8-4(11)7-1/h1H,(H3,5,6,10)(H2,7,8,9,11)/i1+1,7+1

InChI key

POJWUDADGALRAB-SPBYTNOZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling Zhang et al.
American journal of human genetics, 96(4), 597-611 (2015-04-04)
Esophageal squamous cell carcinoma (ESCC) is one of the most common cancers worldwide and the fourth most lethal cancer in China. However, although genomic studies have identified some mutations associated with ESCC, we know little of the mutational processes responsible.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service