Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

607622

Sigma-Aldrich

Fosphenytoin-2,4,5-13C3,15N2 disodium salt heptahydrate

99 atom % 13C, 98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C13H2715N2Na2O7P
Molecular Weight:
441.31
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C
98 atom % 15N

form

solid

mass shift

M+5

SMILES string

O.O.O.O.O.O.O.[Na+].[Na+].[O-]P([O-])(=O)OC[15N]1[13C](=O)[15NH][13C](c2ccccc2)(c3ccccc3)[13C]1=O

InChI

1S/C16H15N2O6P.2Na.7H2O/c19-14-16(12-7-3-1-4-8-12,13-9-5-2-6-10-13)17-15(20)18(14)11-24-25(21,22)23;;;;;;;;;/h1-10H,11H2,(H,17,20)(H2,21,22,23);;;7*1H2/q;2*+1;;;;;;;/p-2/i14+1,15+1,16+1,17+1,18+1;;;;;;;;;

InChI key

KFGQDIBFHJVFDV-WSWAXWECSA-L

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service