Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

607274

Sigma-Aldrich

Carbon-12C dioxide-16O2

99.93 atom % 16O, 99.9 atom % 12C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CO2
Molecular Weight:
44.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

99.9 atom % 12C
99.93 atom % 16O

form

gas

mass shift

depleted

SMILES string

[16O]=[12C]=[16O]

InChI

1S/CO2/c2-1-3/i1+0,2+0,3+0

InChI key

CURLTUGMZLYLDI-SXRZRGFPSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service