Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

606367

Sigma-Aldrich

Bromotrichloromethane-13C

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br13CCl3
Molecular Weight:
199.27
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % D

Quality Level

bp

105 °C (lit.)

mp

-6 °C (lit.)

density

2.022 g/mL at 25 °C

mass shift

M+1

SMILES string

Cl[13C](Cl)(Cl)Br

InChI

1S/CBrCl3/c2-1(3,4)5/i1+1

InChI key

XNNQFQFUQLJSQT-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service