Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

605263

Sigma-Aldrich

Fmoc-Asp-OH-4-13C

98 atom % 13C, 99% (CP)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-aspartic-4-13C acid, L-Aspartic-4-13C acid, N-Fmoc derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO213CCH2CH(NH-Fmoc)CO2H
Molecular Weight:
356.33
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % 13C

assay

99% (CP)

form

solid

optical activity

[α]20/D -13°, c = 1 in DMF

technique(s)

bio NMR: suitable

mp

190 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+1

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](C[13C](O)=O)NC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C19H17NO6/c21-17(22)9-16(18(23)24)20-19(25)26-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15-16H,9-10H2,(H,20,25)(H,21,22)(H,23,24)/t16-/m0/s1/i17+1

InChI key

KSDTXRUIZMTBNV-IBEIMJSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service