Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

604488

Sigma-Aldrich

2-Pentene-2-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH=13CHCH3
Molecular Weight:
71.13
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

description

cis/trans mixture

isotopic purity

99 atom % 13C

mass shift

M+1

SMILES string

CC\C=[13CH]\C

InChI

1S/C5H10/c1-3-5-4-2/h3,5H,4H2,1-2H3/b5-3+/i3+1

InChI key

QMMOXUPEWRXHJS-INSTWSLVSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Press. Gas Liquefied gas - Skin Irrit. 2

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

-49.0 °F - closed cup

flash_point_c

-45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service