Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

604283

Sigma-Aldrich

1-Phenyl-13C6-1-dodecanone

99 atom % 13C

Synonym(s):

Dodecanophen-13C6-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13C6H5-C(O)CH2(CH2)9CH3
Molecular Weight:
266.37
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

mass shift

M+6

SMILES string

CCCCCCCCCCCC(=O)[13c]1[13cH][13cH][13cH][13cH][13cH]1

InChI

1S/C18H28O/c1-2-3-4-5-6-7-8-9-13-16-18(19)17-14-11-10-12-15-17/h10-12,14-15H,2-9,13,16H2,1H3/i10+1,11+1,12+1,14+1,15+1,17+1

InChI key

DJNJZIFFCJTUDS-ITMBUKKYSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service