Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

603783

Sigma-Aldrich

2-(2-Iodoethyl-13C2)-2-methyl-13C-dioxolane-2-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4C2H11IO2
CAS Number:
Molecular Weight:
246.03
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

mass shift

M+4

SMILES string

[13CH3][13C]1([13CH2][13CH2]I)OCCO1

InChI

1S/C6H11IO2/c1-6(2-3-7)8-4-5-9-6/h2-5H2,1H3/i1+1,2+1,3+1,6+1

InChI key

SXVDTZCWOQEFBN-HDPXQERQSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service