Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60020

Sigma-Aldrich

Caffeic acid

purum, ≥95.0% (HPLC)

Synonym(s):

3,4-Dihydroxybenzeneacrylic acid, 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1954563
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥95.0% (HPLC)

form

powder

mp

211-213 °C (dec.) (lit.)

SMILES string

OC(=O)\C=C\c1ccc(O)c(O)c1

InChI

1S/C9H8O4/c10-7-3-1-6(5-8(7)11)2-4-9(12)13/h1-5,10-11H,(H,12,13)/b4-2+

InChI key

QAIPRVGONGVQAS-DUXPYHPUSA-N

Gene Information

human ... ELA2(1991)
rat ... Alox5(25290)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A natural dietary phenolic compound found in plants that is an anti-oxidant. Inhibits the synthesis of leukotrienes that are involved in immunoregulation, inflammation and allergy. Inhibits Cu2+-induced LDL oxidation.
Anti-oxidant phenolic compound found in plants; Inhibits the synthesis of leukotrienes.

Other Notes

Inhibits 5-lipoxygenase activity in mastocyma cells[1]

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Koshihara et al.
Biochimica et biophysica acta, 792(1), 92-97 (1984-01-17)
.eukotrienes are significantly involved in immunoregulation and in a variety of diseases, including asthma, inflammation and various allergic conditions. They are initially biosynthesized by 5-lipoxygenase from arachidonic acid, which can also be metabolized to prostaglandin endoperoxide by cyclooxygenase. The specific
Ren-Wang Jiang et al.
Current medicinal chemistry, 12(2), 237-246 (2005-01-11)
Caffeic acid (3,4-dihydroxycinnamic acid), one of the most common phenolic acids, frequently occurs in fruits, grains and dietary supplements for human consumption as simple esters with quinic acid or saccharides, and are also found in traditional Chinese herbs. Caffeic acid
Jun Liu et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 51, 310-316 (2012-10-27)
The antioxidant activities of ethanolic extract from edible mushroom Agaricus bisporus (A. bisporus) were evaluated by various methods in vitro and in vivo. In antioxidant assays in vitro, ethanolic extract of A. bisporus was found to have strong reducing power
Emmanoel Vilaça Costa et al.
Journal of natural products, 73(6), 1180-1183 (2010-05-19)
Phytochemical investigation of the bark of Guatteria hispida afforded three new alkaloids, 9-methoxy-O-methylmoschatoline (1), 9-methoxyisomoschatoline (2), and isocerasonine (3), along with 10 known alkaloids, 8-oxopseudopalmatine (4), O-methylmoschatoline (5), lysicamine (6), liriodenine (7), 10-methoxyliriodenine (8), nornuciferine (9), anonaine (10), xylopine (11)
Susanne Vogel et al.
Journal of natural products, 71(7), 1237-1241 (2008-07-10)
The minor hop ( Humulus lupulus) chalcones 3'-geranylchalconaringenin (3), 5'-prenylxanthohumol (4), flavokawin (5), xanthohumol H (8), xanthohumol C (9), and 1'',2''-dihydroxanthohumol C (10) were synthesized. The non-natural chalcones 3'-geranyl-6'-O-methylchalconaringenin (2), 3'-methylflavokawin (6), and 2'-O-methyl-3'-prenylchalconaringenin (7) were also synthesized. Cytotoxicity was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service