Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

597716

Sigma-Aldrich

4,5-Dichloro-2-methyl-3(2H)-pyridazinone

97%

Synonym(s):

4,5-Dichloro-2-methylpyridazin-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4Cl2N2O
CAS Number:
Molecular Weight:
179.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

87-91 °C (lit.)

SMILES string

CN1N=CC(Cl)=C(Cl)C1=O

InChI

1S/C5H4Cl2N2O/c1-9-5(10)4(7)3(6)2-8-9/h2H,1H3

InChI key

ACKBTCUMGAHRIE-UHFFFAOYSA-N

Application

4,5-Dichloro-2-methyl-3(2H)-pyridazinone can be used in the synthesis of 4-amino-3(2H)-pyridazinones and 5-amino-3(2H)-pyridazinones via reaction with N-benzylaminoethanol.[1]

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Lipophilicity of regioisomers: a case study on 3 (2H)-pyridazinones"
<BIG>Karolyhazy L, et al.</BIG>
Journal of Molecular Structure: THEOCHEM, 578(01), 89-91 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service