Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

597473

Sigma-Aldrich

4-(2-Bromoethoxy)-3-methoxybenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2OC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
259.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

66-70 °C (lit.)

functional group

aldehyde
bromo

SMILES string

[H]C(=O)c1ccc(OCCBr)c(OC)c1

InChI

1S/C10H11BrO3/c1-13-10-6-8(7-12)2-3-9(10)14-5-4-11/h2-3,6-7H,4-5H2,1H3

InChI key

VFSWCDPTQADMSM-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service