Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

597309

Sigma-Aldrich

PAMAM dendrimer, 1,12-diaminododecane core, generation 0.0 solution

20 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)12NH2]:(G=0);dendri PAMAM(NH2)4
Molecular Weight:
656.95
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

description

amino surface groups

form

liquid

mol wt

generation 0

feature

core type 1,12-diaminododecane core (12-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

4

refractive index

n20/D 1.367

bp

64.7 °C

density

0.791 g/mL at 25 °C

storage temp.

−20°C

SMILES string

NCCNC(=O)CCN(CCCCCCCCCCCCN(CCC(=O)NCCN)CCC(=O)NCCN)CCC(=O)NCCN

InChI

1S/C32H68N10O4/c33-15-19-37-29(43)11-25-41(26-12-30(44)38-20-16-34)23-9-7-5-3-1-2-4-6-8-10-24-42(27-13-31(45)39-21-17-35)28-14-32(46)40-22-18-36/h1-28,33-36H2,(H,37,43)(H,38,44)(H,39,45)(H,40,46)

InChI key

MVPBOIZWNUVCLB-UHFFFAOYSA-N

Features and Benefits

Contains 4 surface amino groups.

Packaging

unit weight includes solvent

Legal Information

Mfg. by Dendritic Nanotechnologies Inc. Licensed for research use only.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service