Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

596426

Sigma-Aldrich

PAMAM dendrimer, 1,4-diaminobutane core, generation 6.0 solution

10 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)4NH2]:(G=6);dendri PAMAM(NH2)256
Molecular Weight:
58074.17
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amino surface groups

form

solution

mol wt

generation 6

feature

core type 1,4-diaminobutane core (4-carbon core)

concentration

10 wt. % in methanol

no. Surface Groups

256

refractive index

n20/D 1.347

bp

64.7 °C

density

0.791 g/mL at 25 °C

storage temp.

−20°C

Features and Benefits

Contains 256 surface amino groups.

Packaging

unit weight includes solvent

Legal Information

Mfg. by Dendritic Nanotechnologies Inc. Licensed for research use only.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service