Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

594563

Sigma-Aldrich

He-3/Ne-20/Ne-22 Gas Mixture

ratio (44:3:3), 99.9 atom % 22Ne, 99.95 atom % 20Ne

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12142200

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99.95 atom % 20Ne
99.9 atom % 22Ne

form

gas

composition

ratio (44:3:3)

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service