Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

594512

Sigma-Aldrich

2-(Trifluoromethyl)benzamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6F3NO
CAS Number:
Molecular Weight:
189.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

160-164 °C (lit.)

SMILES string

NC(=O)c1ccccc1C(F)(F)F

InChI

1S/C8H6F3NO/c9-8(10,11)6-4-2-1-3-5(6)7(12)13/h1-4H,(H2,12,13)

InChI key

QBAYIBZITZBSFO-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Rouquié et al.
Regulatory toxicology and pharmacology : RTP, 70(3), 673-680 (2014-12-03)
Fluopyram, a broad spectrum fungicide, caused an increased incidence of thyroid follicular cell (TFC) adenomas in males at the highest dose evaluated (750ppm equating to 105mg/kg/day) in the mouse oncogenicity study. A series of short-term mechanistic studies were conducted in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service