Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

59436

Sigma-Aldrich

Methyl (4R,5S)-2,2,5-trimethyl-1,3-dioxolane-4-carboxylate

technical, ≥90% (sum of enantiomers, GC)

Synonym(s):

4-Deoxy-2,3-O-isopropylidene-L-threonic acid methyl ester, Methyl (2R,3S)-2,3-O-isopropylidene-2,3-dihydroxybutyrate, Methyl 4-deoxy-2,3-O-isopropylidene-L-threonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
Beilstein/REAXYS Number:
4667377
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

grade

technical

assay

≥90% (sum of enantiomers, GC)

optical activity

[α]20/D +19±1°, c = 4% in chloroform

refractive index

n20/D 1.422

density

1.06 g/mL at 20 °C (lit.)

SMILES string

COC(=O)[C@@H]1OC(C)(C)O[C@H]1C

InChI

1S/C8H14O4/c1-5-6(7(9)10-4)12-8(2,3)11-5/h5-6H,1-4H3/t5-,6+/m0/s1

InChI key

AELKYRWKHKGMAD-NTSWFWBYSA-N

Other Notes

Chiral building block: stereoselective α-alkylation.[1] Preparation of the corresponding aldehyde.[2][3]

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

122.0 °F - closed cup

flash_point_c

50 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Fronza et al.
Tetrahedron Letters, 3883-3883 (1979)
W. Ladner
Chemische Berichte, 116, 3413-3413 (1983)
S. Servi
The Journal of Organic Chemistry, 50, 5865-5865 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service