Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

591734

Sigma-Aldrich

(3R)-(−)-3-(methyl-1H-indol-3-yl)-3-phenylpropionic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO2
CAS Number:
Molecular Weight:
279.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

assay

97%

optical activity

[α]20/D -44°, c = 1% in chloroform

mp

135-139 °C

SMILES string

Cn1cc([C@H](CC(O)=O)c2ccccc2)c3ccccc13

InChI

1S/C18H17NO2/c1-19-12-16(14-9-5-6-10-17(14)19)15(11-18(20)21)13-7-3-2-4-8-13/h2-10,12,15H,11H2,1H3,(H,20,21)/t15-/m1/s1

InChI key

XVWRQAZBQVTVJA-OAHLLOKOSA-N

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service