Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

591300

4-Oxo-TEMPO-1-15N

98 atom % 15N

Synonym(s):

4-Oxo-2,2,6,6-tetramethylpiperidine-1-15N-1-oxyl, TEMPONE-1-15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H1615NO2
Molecular Weight:
171.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

isotopic purity

98 atom % 15N

mp

35 °C (lit.)

mass shift

M+1

SMILES string

CC1(C)CC(=O)CC(C)(C)[15N]1[O]

InChI

1S/C9H16NO2/c1-8(2)5-7(11)6-9(3,4)10(8)12/h5-6H2,1-4H3/i10+1

InChI key

WSGDRFHJFJRSFY-DETAZLGJSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service