Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59110

Sigma-Aldrich

Isopentylbenzene

≥97.0%

Synonym(s):

(3-Methyl-1-butyl)benzene, Isoamylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
148.24
Beilstein/REAXYS Number:
1855612
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0%

refractive index

n20/D 1.486

bp

197 °C (lit.)

density

0.854 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)CCc1ccccc1

InChI

1S/C11H16/c1-10(2)8-9-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3

InChI key

XNXIYYFOYIUJIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

149.0 °F - closed cup

flash_point_c

65 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura A Mertens et al.
The journal of physical chemistry. A, 122(24), 5418-5436 (2018-05-09)
The relative rates of C-C and C-H β-scission reactions of isobutyl radicals (2-methylprop-1-yl, C4H9) were investigated with shock tube experiments at temperatures of (950 to 1250) K and pressures of (200 to 400) kPa. We produced isobutyl radicals from the
Veronika Pilařová et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1043, 25-32 (2016-09-22)
The new microextraction technique named parallel artificial liquid membrane extraction (PALME) was introduced as an alternative approach to liquid-liquid extraction of charged analytes from aqueous samples. The concept is based on extraction of analytes across a supported liquid membrane sustained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service