Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

590576

Sigma-Aldrich

PAMAM-OS-trimethoxysilyl dendrimer, generation 6 solution

5 wt. % in isopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=6);dendri PAMAM((NHCH2CH2CO2(CH2)3Si(OCH3)3)512
Molecular Weight:
178018.83
MDL number:
UNSPSC Code:
12162002

description

trimethoxysilyl surface group

form

liquid

mol wt

generation 6

feature

core type ethylenediamine core (2-carbon core)

concentration

5 wt. % in isopropanol

no. Surface Groups

512

storage temp.

2-8°C

Features and Benefits

Contains 512 surface organosilicon groups.

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service