Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

590525

Sigma-Aldrich

O-Methylisourea-d6 deuteriochloride

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3OC(=ND)ND2 · DCl
CAS Number:
Molecular Weight:
117.59
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

form

solid

mp

115 °C (lit.)

mass shift

M+7

SMILES string

Cl[2H].[2H]\N=C(\OC([2H])([2H])[2H])N([2H])[2H]

InChI

1S/C2H6N2O.ClH/c1-5-2(3)4;/h1H3,(H3,3,4);1H/i1D3;/hD4

Inchi Key

MUDVUWOLBJRUGF-KYWLLVRESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service