Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

590223

Sigma-Aldrich

Sodium ruthenate, polymer-bound FibreCat®

Synonym(s):

Sodium perruthenate, polymer-bound FibreCat®

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

Application

Used as a recyclable supported catalyst for the selective oxidation of alcohols to aldehydes with very little over oxidation to the acid.

Legal Information

FibreCat is a registered trademark of Johnson Matthey PLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service