Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59020

Sigma-Aldrich

Octadecanol – hexadecanol – eicosanol mixture (regio isomeric mixture)

≥95.0% (GC)

Synonym(s):

Fatty alcohol mixture (C16, C18, C20)

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein/REAXYS Number:
1703362
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥95.0% (GC)

refractive index

n20/D 1.451

functional group

hydroxyl

SMILES string

CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCCCO

InChI

1S/C20H42O.C18H38O.C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19;1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h21H,2-20H2,1H3;19H,2-18H2,1H3;17H,2-16H2,1H3

InChI key

VPOYYFMLXVUTRQ-UHFFFAOYSA-N

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juergen Poerschmann et al.
The Science of the total environment, 578, 219-227 (2016-02-06)
Natural neutralization of acidic mining lakes is often limited by organic matter. The knowledge of the sources and degradability of organic matter is crucial for understanding alkalinity generation in these lakes. Sediments collected at different depths (surface sediment layer from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service