Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

589217

Sigma-Aldrich

Sodium L-lactate-2-d1 solution

98 atom % D, 45-55 % (w/w) in H2O

Synonym(s):

L-Lactic-2-d1 acid sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CD(OH)CO2Na
Molecular Weight:
113.07
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

description

Packaged as gram of solution, 45-55% (w/w)

isotopic purity

98 atom % D

optical activity

[α]25/D -12°, c = 1 in H2O

concentration

45-55 % (w/w) in H2O

mp

163-165 °C (lit.)

mass shift

M+1

storage temp.

−20°C

SMILES string

[Na+].[2H][C@@](C)(O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i2D;

InChI key

NGSFWBMYFKHRBD-QEKRQAICSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service