Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

589195

1-Methyl-1-propanethiol-d10

98 atom % D

Synonym(s):

2-Butanethiol-d10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD(SD)CD2CD3
Molecular Weight:
100.25
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

98 atom % D

bp

85 °C (lit.)

density

0.921 g/mL at 25 °C

mass shift

M+10

SMILES string

[2H]SC([2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C4H10S/c1-3-4(2)5/h4-5H,3H2,1-2H3/i1D3,2D3,3D2,4D/hD

InChI key

LOCHFZBWPCLPAN-LPIDSFCWSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service