Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

589160

Sigma-Aldrich

Poly(2-propylacrylic acid)

average Mn ~100,000 by GPC

Synonym(s):

α-Propylacrylic acid homopolymer, 2-Propylacrylic acid homopolymer, Polypropylacrylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn ~100,000 by GPC

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

pH-sensitive polymer for drug delivery.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service