Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

589136

Sigma-Aldrich

Benzyl 2-ethyl acrylate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(C2H5)CO2CH2C6H5
Molecular Weight:
190.24
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

99%

storage temp.

−20°C

SMILES string

CCC(=C)C(=O)OCc1ccccc1

InChI

1S/C12H14O2/c1-3-10(2)12(13)14-9-11-7-5-4-6-8-11/h4-8H,2-3,9H2,1H3

InChI key

DGMZHLOQOORCIE-UHFFFAOYSA-N

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service