Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

588431

Sigma-Aldrich

Poly(ethylene oxide), 6-arm, anthracene terminated

average Mn ~10,000

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

mol wt

Mw/Mn <1.2
average Mn ~10,000

mp

48-53 °C (lit.)

fluorescence

λex 251 nm; λem 458 nm in chloroform

Application

Photocrosslinkable PEG-based hydrophilic biomaterial.

Features and Benefits

Undergoes reversible photocrosslinking to a hydrogel with tunable physicochemical properties including swellability, absorption spectrum, and topography.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng, Y. et al.
Macromolecules, 35, 5228-5234 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service