Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

588377

Sigma-Aldrich

5-Bromopentanoic-3,3,4,4-d4 acid

≥99 atom % D, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2(CD2)2CH2COOH
CAS Number:
Molecular Weight:
185.05
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥99 atom % D

assay

99% (CP)

form

solid

mp

38-40 °C (lit.)

mass shift

M+4

SMILES string

OC(CC([2H])([2H])C([2H])([2H])CBr)=O

InChI

1S/C5H9BrO2/c6-4-2-1-3-5(7)8/h1-4H2,(H,7,8)/i1D2,2D2

InChI key

WNXNUPJZWYOKMW-LNLMKGTHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service