Sign In to View Organizational & Contract Pricing.
About This Item
Linear Formula:
Ag2WO4
CAS Number:
Molecular Weight:
463.57
UNSPSC Code:
12352300
PubChem Substance ID:
EC Number:
236-708-3
MDL number:
Skip To
InChI key
QEKREONBSFPWTQ-UHFFFAOYSA-N
InChI
1S/2Ag.4O.W/q2*+1;;;2*-1;
SMILES string
[Ag+].[Ag+].[O-][W]([O-])(=O)=O
assay
99.95% trace metals basis
form
powder
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service