Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

579491

Sigma-Aldrich

2,3-Difluorobenzamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5F2NO
CAS Number:
Molecular Weight:
157.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

114-119 °C (lit.)

SMILES string

NC(=O)c1cccc(F)c1F

InChI

1S/C7H5F2NO/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3H,(H2,10,11)

InChI key

DDAINDMVKSETGF-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula V Gonzalez et al.
Parasites & vectors, 13(1), 257-257 (2020-05-18)
Aedes aegypti (L.) is the main vector of dengue, yellow fever, Zika and chikungunya viruses. A new method for controlling this mosquito has been developed based on the possibility that wild adult mosquitoes exposed to artificial resting sites contaminated with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service