Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

575038

Sigma-Aldrich

Potassium hexaiodorhenate(IV)

>99.99% trace metals basis

Synonym(s):

Potassium hexaiodorhenate, Potassium hexaiodorhenate(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2ReI6
CAS Number:
Molecular Weight:
1025.83
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
>99.99% trace metals basis
form:
powder

assay

>99.99% trace metals basis

form

powder

impurities

≤200 ppm Trace Metal Impurities Analysis

SMILES string

[K+].[K+].I[Re--](I)(I)(I)(I)I

InChI

1S/6HI.2K.Re/h6*1H;;;/q;;;;;;2*+1;+4/p-6

InChI key

DFWHBRZEYYLAED-UHFFFAOYSA-H

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service