Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

574198

Sigma-Aldrich

Silver(I) perrhenate

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgReO4
CAS Number:
Molecular Weight:
358.07
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99.99% trace metals basis
form:
powder

assay

99.99% trace metals basis

form

powder

impurities

≤150 ppm Trace Metal Analysis

mp

430 °C (lit.)

density

7.05 g/mL at 25 °C (lit.)

SMILES string

[Ag+].[O-][Re](=O)(=O)=O

InChI

1S/Ag.4O.Re/q+1;;;;-1;

InChI key

JRISYOCHUNKJAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service