Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

573876

Sigma-Aldrich

Ammonium hydrogenoxalate hydrate

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)HC2O4 · xH2O
Molecular Weight:
107.07 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

assay

99.999% trace metals basis

form

solid

impurities

≤20 ppm Trace metallic impurities

mp

210 °C (dec.) (lit.)

SMILES string

N.O.OC(=O)C(O)=O

InChI

1S/C2H2O4.H3N.H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);1H3;1H2

InChI key

JRWKBXBBNURFHV-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service