Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

572977

Sigma-Aldrich

Benzenesulfonyl azide, polymer-bound

macroporous, 50-100 mesh, extent of labeling: ~1.5 mmol/g loading

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

~1.5 mmol/g loading

particle size

50-100 mesh

Application

Polymer-supported diazo transfer agent.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polystyrene-supported benzenesulfonyl azide: a diazo transfer reagent that is both efficient and safe.
G M Green et al.
The Journal of organic chemistry, 66(7), 2509-2511 (2001-04-03)

Articles

Since the preparation of the first organic azide, phenyl azide, by Peter Griess in 1864 this energy-rich and versatile class of compounds has enjoyed considerable interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service