Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

572640

Sigma-Aldrich

4-Methylbenzhydryl chloride, polymer-bound

100-200 mesh, extent of labeling: 1.6-2.2 mmol/g loading, 1 % cross-linked with divinylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reactivity: nucleophile reactive

extent of labeling

1.6-2.2 mmol/g loading

particle size

100-200 mesh

Application

The benzyhydryl chloride resins are used for the attachment of amines, alcohols, sulfamates, thiols and phenols using the Fmoc strategy. These resins are an intermediate between the Merrifield resin and the trityl chloride resins in term of acid lability. The carbocation formed during the cleavage reaction is less stable than the Merrifield resin but more stable than the trityl chloride resin. The 4-Merthylbenzhydryl chloride resin is a more acid labile resin than the benzhydryl chloride resin.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service