572179
cis-1-Propen-1-ylboronic acid
≥95.0%
Synonym(s):
cis-Propenylboronic acid
About This Item
Recommended Products
assay
≥95.0%
composition
trans isomer, ≤10%
mp
65-70 °C (lit.)
storage temp.
−20°C
SMILES string
C\C=C/B(O)O
InChI
1S/C3H7BO2/c1-2-3-4(5)6/h2-3,5-6H,1H3/b3-2-
InChI key
CBMCZKMIOZYAHS-IHWYPQMZSA-N
Application
- As an intermediate in the preparation of furan-2-yl analog of salvinorin A, a psychoactive natural product.[1]
- To synthesize antibacterial polyoxygenated dibenzofuran derivatives from phloroglucinol.[2]
- As a substrate in the palladium-catalyzed cis-vinyl arenes synthesis by reacting with various aryl chlorides.[3]
Other Notes
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
219.2 °F - closed cup
flash_point_c
104.00 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service