Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

571660

Sigma-Aldrich

Poly(9,9-di-n-dodecylfluorenyl-2,7-diyl)

Synonym(s):

PFD, Poly(9,9-di-n-dodecylfluorene)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

solubility

chloroform: soluble
methylene chloride: soluble
toluene: soluble

λmax

≤369 nm

SMILES string

CCCCCCCCCCCCC1(CCCCCCCCCCCC)c2ccccc2-c3ccccc13

Looking for similar products? Visit Product Comparison Guide

General description

Conjugated polymer. Exhibits blue photo- and electroluminescence.
Poly(9,9-di-n-dodecylfluorenyl-2,7-diyl) (PFDD) is a polyfluorene based conjugated polymer with thermotropic liquid crystallinity, that can be used as a blue-light emitting polymer in organic electronics.[1][2]

application

PFDD is used as a semiconducting polymer for use in the development of organic thin film transistors (OTFTs) and organic light emitting diodes(OLEDs). It forms a composite with carbon nanomaterials for obtaining high tensile films with improved conductivity.[3][4]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of temperature on the selection of semiconducting single walled carbon nanotubes using Poly (3-dodecylthiophene-2, 5-diyl)
Gomulya W, et al.
Carbon, 84(4), 66-73 (2015)
Liquid crystalline polyfluorenes for blue polarized electroluminescence
Macromolecular Symposia, 154(1), 139-148 (2000)
Enrichment of large-diameter semiconducting SWCNTs by polyfluorene extraction for high network density thin film transistors
Ding J, et al.
Nanoscale, 6(4), 2328-2339 (2014)
Carbon nanotube network ambipolar field-effect transistors with 108 on/off ratio
Derenskyi V, et al.
Advanced Materials, 26(34), 5969-5975 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service