Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

57135

Sigma-Aldrich

Indium(III) nitrate hydrate

technical, 32-36% indium basis

Synonym(s):

Indium trinitrate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In(NO3)3 · aq
CAS Number:
Molecular Weight:
300.83 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

technical

reaction suitability

reagent type: catalyst
core: indium

concentration

32-36% In

impurities

20-25% water

SMILES string

O.[In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/In.3NO3.H2O/c;3*2-1(3)4;/h;;;;1H2/q+3;3*-1;

InChI key

YZZFBYAKINKKFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame over circleExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Ox. Sol. 2

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new weapon in the treatment of non-Hodgkin's lymphoma.
James D Lipcamon et al.
Radiology management, 24(5), 13-15 (2002-11-09)
Eun-Sil Kang et al.
Journal of nanoscience and nanotechnology, 7(11), 4053-4056 (2007-12-01)
InGaN/GaN multiple quantum wells (MQWs) were successfully grown on the inclined GaN(1101) microfacets. Conventional photolithography and subsequent growth of GaN were employed to generate the V-shaped microfacets along (1120) direction. The well-developed microfacets observed by scanning electron microscopy and the
V Brix-Christensen et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 112(2), 133-140 (2004-04-02)
The aim was to investigate if organ dysfunction is a consequence of cell accumulation in the tissue and whether this accumulation is caused by the cardiopulmonary bypass (CPB) procedure. Twenty-six piglets were used in the sham group (sternotomy, n=12) or
Tingjiang Yan et al.
Journal of colloid and interface science, 325(2), 425-431 (2008-06-17)
Nearly monodisperse single-crystalline In(OH)(3) nanocubes were successfully synthesized using In(NO(3))(3) x 4.5 H(2)O as indium source in the presence of urea and cetyltrimethyl ammonium bromide (CTAB) by a two-step hydrothermal process: the stock solution was heated at 70 degrees C
Jorge L Zurita et al.
The Science of the total environment, 387(1-3), 155-165 (2007-09-07)
Indium nitrate is mainly used as a semiconductor in batteries, for plating and other chemical and medical applications. There is a lack of available information about the adverse effects of indium compounds on aquatic organisms. Therefore, the toxic effects on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service