570478
Poly[(methyl methacrylate)-co-(Disperse Red 13 acrylate)]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
transition temp
Tg 94 °C
solubility
chloroform: soluble
toluene: soluble
λmax
492 nm
General description
Chromophore monomer ca. 25 mol %.
Application
NLO polymer
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service