Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

570346

Sigma-Aldrich

Poly(ethylene oxide)-block-polycaprolactone, 4-arm

PEG average Mn ~2,500

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

PCL average Mn ~2,500
PEG average Mn ~2,500
average Mn ~5,000 (total)

degradation timeframe

>12 months

mp

53-55 °C (lit.)

Mw/Mn

<1.2

Features and Benefits

Biocompatible polymer

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Hydrogel-based biomaterials for cell delivery and tissue regeneration applications are discussed.

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service