570265
Poly{[2-[2′,5′-bis(2″-ethylhexyloxy)phenyl]-1,4-phenylenevinylene]-co-[2-methoxy-5-(2′-ethylhexyloxy)-1,4-phenylenevinylene]}
Synonym(s):
BEHP-co-MEH-PPV, Poly[(BEHP-PPV)-co-(MEH-PPV)]
About This Item
Recommended Products
form
solid
mol wt
Mn 18,900 (typical)
composition
BEHP-PPV, ~60 mol %
MEH-PPV, ~40 mol %
solubility
THF: soluble
chloroform: soluble
xylene: soluble
fluorescence
λex 325 nm; λem 545 nm in chloroform
Mw/Mn
4.7 (typical)
General description
Application
Features and Benefits
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service