Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

570168

Sigma-Aldrich

2-Ethoxycarbonylphenylboronic acid pinacol ester

97%

Synonym(s):

2-Carboethoxyphenylboronic acid pinacol ester, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4COOCH2CH3BO2C2(CH3)4
CAS Number:
Molecular Weight:
276.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

60-65 °C

functional group

ester

SMILES string

CCOC(=O)c1ccccc1B2OC(C)(C)C(C)(C)O2

InChI

1S/C15H21BO4/c1-6-18-13(17)11-9-7-8-10-12(11)16-19-14(2,3)15(4,5)20-16/h7-10H,6H2,1-5H3

InChI key

RFBZWPFBCXBBJS-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service