569917
(R)-(+)-5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′-2-naphthol
95%
About This Item
Recommended Products
assay
95%
form
solid
optical activity
[α]20/D +73°, c = 1 in THF
mp
160-166 °C (lit.)
InChI
1S/C20H22O2/c21-17-11-9-13-5-1-3-7-15(13)19(17)20-16-8-4-2-6-14(16)10-12-18(20)22/h9-12,21-22H,1-8H2
InChI key
UTXIFKBYNJRJPH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- As a starting material for the preparation of octahydro-binaphthyl based organic light-emitting diodes.[1]
- As a ligand in the 1,4-hydrophosphinylation of methyl cyclohexene-1-carboxylate using diphenylphosphine oxide, applicable in the synthesis of CyPHOX ligands.[2]
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service