Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

569895

Sigma-Aldrich

Potassium permanganate on silica gel

phosphate buffered, 200-400 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KMnO4-silica
MDL number:
UNSPSC Code:
12352200

form

solid

particle size

200-400 mesh

capacity

15-20 %, silica gel

SMILES string

[K+].O=[Si]=O.[O-][Mn](=O)(=O)=O

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

target_organs

Brain

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service