568767
Chlorite, polymer-bound
Macroporous, 20-50 mesh, extent of labeling: ~2.0 mmol/g loading
Synonym(s):
Chlorite on Amberlite™ IRA-900
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
solid
reaction suitability
reaction type: solution phase peptide synthesis
extent of labeling
~2.0 mmol/g loading
particle size
20-50 mesh
SMILES string
[O-]Cl=O.C[N+](C)(C)C.C=Cc1ccccc1.C=Cc2ccc(C=C)cc2
Application
Polymer-supported oxidizing reagent.
Legal Information
Amberlite is a trademark of DuPont de Nemours, Inc.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Takemoto, T.
Synlett, 10, 1555-1555 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service