Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

568007

Sigma-Aldrich

Dimethyldiphenylsilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2Si(CH3)2
CAS Number:
Molecular Weight:
212.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.5640 (lit.)

bp

276 °C/740 mmHg (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(c1ccccc1)c2ccccc2

InChI

1S/C14H16Si/c1-15(2,13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12H,1-2H3

InChI key

WJKVFIFBAASZJX-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

221.0 °F - closed cup

flash_point_c

105.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service