Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

567876

Sigma-Aldrich

4-Ethylphenethyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C6H4CH2CH2NCO
CAS Number:
Molecular Weight:
175.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.5150 (lit.)

bp

225-226 °C (lit.)

density

1.008 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCc1ccc(CCN=C=O)cc1

InChI

1S/C11H13NO/c1-2-10-3-5-11(6-4-10)7-8-12-9-13/h3-6H,2,7-8H2,1H3

InChI key

RBYBLAVTEAMCBH-UHFFFAOYSA-N

Caution

Refrigerate

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

170.1 °F - closed cup

flash_point_c

76.7 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service