566098
TEMPO, polymer-bound
100-200 mesh, extent of labeling: 1.0 mmol/g loading, 1 % cross-linked with divinylbenzene
Synonym(s):
(2,2,6,6-Tetramethyl-1-piperidinyloxy, free radical), polymer-bound, PS-TEMPO
About This Item
Recommended Products
form
solid
crosslinking
1 % cross-linked with divinylbenzene
reaction suitability
reaction type: solution phase peptide synthesis
reactivity: alcohol reactive
extent of labeling
1.0 mmol/g loading
particle size
100-200 mesh
General description
Features and Benefits
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service